3-Chloro-2-methoxy-5-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-methoxypyridine-5-boronic acid

A166737-1G A166737-250MG A166737-5G
76985-240EA 103.97 USD
76985-240 76985-242 76985-244
3-Chloro-2-methoxy-5-pyridineboronic acid 98%
3-Chloro-2-methoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
MDL Number: MFCD04972386
CAS Number: 942438-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR