MI-2 ≥98%

Supplier: Ambeed
Warning

Synonyms: Menin-MLL inhibitor 2, 4-[4-(5,5-dimethyl-4H-1,3-thiazol-2-yl)piperazin-1-yl]-6-propylthieno[2,3-d]pyrimidine, Menin-MLL inhibitor MI-2, 4-(4-(5,5-Dimethyl-4,5-dihydrothiazol-2-yl)piperazin-1-yl)-6-propylthieno[2,3-d]pyrimidine

A166598-10MG A166598-1MG A166598-2MG A166598-50MG A166598-5MG A166598-100MG A166598-1G A166598-250MG
76985-230EA 28.65 USD
76985-230 76985-232 76985-234 76985-236 76985-238 77683-364 77683-365 77683-366
MI-2 ≥98%
MI-2

Precaución: For research use only.

Formula: C₁₈H₂₅N₅S₂
MW: 375.55 g/mol
MDL Number: MFCD22575021
CAS Number: 1271738-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR