Guanine 95% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 2-Amino-1H-purin-6(7H)-one

A166483-100G A166483-500G A166483-25G A166483-1000G
77011-420EA 20 USD
77011-420 77011-422 77011-424 77320-588 77890-082
Guanine 95% (by HPLC)
Guanine

Precaución: for Research Use Only

Formula: C₅H₅N₅O
MW: 151.13 g/mol
MDL Number: MFCD00071533
CAS Number: 73-40-5
Merck Index: 13,04580

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR