2-Chloro-N,6-dimethylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A166420-100MG A166420-1G A166420-5G
77011-414EA 12.74 USD
77011-414 77011-416 77011-418
2-Chloro-N,6-dimethylpyrimidin-4-amine 95%
2-Chloro-N,6-dimethylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MW: 157.60 g/mol
MDL Number: MFCD09864578
CAS Number: 3569-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR