3,3'-Thiodipropionic acid 99%

Supplier: Ambeed
Warning

Synonyms: 3,3-Thiodipropionic acid

A166235-1000G A166235-100G A166235-25G A166235-500G
76998-032EA 50.93 USD
76998-032 76998-034 76998-036 76998-038
3,3'-Thiodipropionic acid 99%
3,3'-Thiodipropionic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₄S
MW: 178.21 g/mol
MDL Number: MFCD00002781
CAS Number: 111-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR