(6-(Benzyloxy)-2,3-difluorophenyl)boronic acid 97%

Supplier: Ambeed
Warning

A166070-1G A166070-250MG A166070-5G
77028-858EA 40 USD
77028-858 77028-860 77871-834
(6-(Benzyloxy)-2,3-difluorophenyl)boronic acid 97%
(6-(Benzyloxy)-2,3-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
MDL Number: MFCD12026697
CAS Number: 1451393-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR