3-Fluoro-2-nitrobenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-2-nitrophenyl)methanol

A166056-1G A166056-250MG A166056-5G
77011-410EA 154.9 USD
77011-410 77011-412 77683-361
3-Fluoro-2-nitrobenzyl alcohol 95%
3-Fluoro-2-nitrobenzyl alcohol

Precaución: For research use only.

Formula: C₇H₆FNO₃
MW: 171.126 g/mol
MDL Number: MFCD13185620
CAS Number: 1214323-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR