1-Phenyl-1H-pyrazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A166041-5G A166041-1G A166041-250MG
77011-408EA 81 USD
77011-408 77890-080 77890-081
1-Phenyl-1H-pyrazole-4-carboxylic acid 95%
1-Phenyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00514518
CAS Number: 1134-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR