2-Iodopyridin-3-amine 96%

Supplier: Ambeed
Warning

A165985-10G A165985-250MG A165985-25G A165985-5G A165985-1G
77011-402EA 420.12 USD
77011-402 77011-404 77011-406 77611-616 77683-360
2-Iodopyridin-3-amine 96%
2-Iodopyridin-3-amine

Precaución: For research use only.

Formula: C₅H₅IN₂
MW: 220.0100 g/mol
MDL Number: MFCD07779506
CAS Number: 209286-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR