1-Propene 1,3-Sultone 98%

Supplier: Ambeed
Warning

Synonyms: Prop-1-ene-1,3-sultone, 5H-1,2-Oxathiole 2,2-Dioxide

A165932-10G A165932-1G A165932-250MG A165932-25G A165932-5G
77028-820EA 11.67 USD
77028-820 77028-822 77028-824 77028-826 77028-828
1-Propene 1,3-Sultone 98%
1-Propene 1,3-Sultone

Precaución: for Research Use Only

Formula: C₃H₄O₃S
MW: 120.12 g/mol
MDL Number: MFCD12405143
CAS Number: 21806-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR