5-Methoxy-1H-indazole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A165871-100MG A165871-1G A165871-250MG
77011-390EA 43 USD
77011-390 77011-392 77011-394
5-Methoxy-1H-indazole-3-carbaldehyde 95%
5-Methoxy-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD06738288
CAS Number: 169789-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR