4-Fluoro-1H-indazol-6-amine 95%

Supplier: Ambeed
Warning

A165624-100MG A165624-1G A165624-250MG
77011-384EA 56 USD
77011-384 77011-386 77011-388
4-Fluoro-1H-indazol-6-amine 95%
4-Fluoro-1H-indazol-6-amine

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MDL Number: MFCD07781401
CAS Number: 885520-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR