1-Ethyl-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A165616-100MG A165616-1G A165616-250MG A165616-5G
77028-812EA 13 USD
77028-812 77028-814 77028-816 77028-818
1-Ethyl-1H-pyrazole-4-carboxylic acid 97%
1-Ethyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD02090883
CAS Number: 400858-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR