3-(2-(tert-Butoxycarbonyl)hydrazinyl)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
A165551-1G
A165551-250MG
A165551-5G
77011-378EA
58.35
USD
77011-378
77011-380
77683-358
3-(2-(tert-Butoxycarbonyl)hydrazinyl)benzoic acid 97%
3-(2-(tert-Butoxycarbonyl)hydrazinyl)benzoic acid
Precaución: For research use only.
Formula:
C₁₂H₁₆N₂O₄ MW: 252.26 g/mol |
MDL Number:
MFCD06658360 CAS Number: 156899-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...