2-Amino-3-fluoropyridine 98%

Supplier: Ambeed
Warning

A165540-100G A165540-10G A165540-1G A165540-25G A165540-5G A165540-250MG
76998-018EA 265 USD
76998-018 76998-020 76998-022 76998-024 76998-026 77600-334
2-Amino-3-fluoropyridine 98%
2-Amino-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅FN₂
MDL Number: MFCD04114135
CAS Number: 21717-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR