2-(2-(2-Hydroxyethoxy)ethoxy)ethyl 4-methylbenzenesulfonate 95%
Supplier: Ambeed
|
Warning
|
A165302-100MG
A165302-250MG
A165302-5G
A165302-25G
A165302-1G
77011-368EA
8
USD
77011-368
77011-370
77011-372
77611-608
77890-079
2-(2-(2-Hydroxyethoxy)ethoxy)ethyl 4-methylbenzenesulfonate 95%
2-(2-(2-Hydroxyethoxy)ethoxy)ethyl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₃H₂₀O₆S |
MDL Number:
MFCD18433414 CAS Number: 77544-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...