7-Benzylamino-4-nitrobenz-2-oxa-1,3-diazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Benzylamino-7-nitrobenzofurazan, N-Benzyl-7-nitrobenzo[c][1,2,5]oxadiazol-4-amine
A165132-1MG
A165132-25MG
76998-004EA
5.31
USD
76998-004
76998-006
7-Benzylamino-4-nitrobenz-2-oxa-1,3-diazole 97%
7-Benzylamino-4-nitrobenz-2-oxa-1,3-diazole
Precaución: for Research Use Only
Formula:
C₁₃H₁₀N₄O₃ MW: 270.24 g/mol |
MDL Number:
MFCD00038006 CAS Number: 18378-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...