tert-Butyl 5-chlorospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

Supplier: Ambeed
Warning

A165090-100MG A165090-1G A165090-250MG
77028-784EA 146.41 USD
77028-784 77028-786 77028-788
tert-Butyl 5-chlorospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
tert-Butyl 5-chlorospiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₃ClN₂O₂
MW: 322.82 g/mol
MDL Number: MFCD12198568
CAS Number: 637362-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR