(S)-tert-Butyl-1-methyl-2-oxoimidazolidine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-tert-Butyl 1-methyl-2-oxoimidazolidine-4-carboxylate
A165043-1G
A165043-250MG
A165043-10G
A165043-25G
A165043-5G
77028-780EA
10.61
USD
77028-780
77028-782
77667-312
77667-314
77667-316
(S)-tert-Butyl-1-methyl-2-oxoimidazolidine-4-carboxylate 97%
(S)-tert-Butyl-1-methyl-2-oxoimidazolidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₆N₂O₃ MW: 200.24 g/mol |
MDL Number:
MFCD09952527 CAS Number: 83056-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...