Methyl 4-ethyl-3-iodobenzoate 95%

Supplier: Ambeed
Warning

A164869-250MG A164869-100MG A164869-1G
76985-210EA 98 USD
76985-210 77890-077 77890-078
Methyl 4-ethyl-3-iodobenzoate 95%
Methyl 4-ethyl-3-iodobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁IO₂
MDL Number: MFCD11112907
CAS Number: 51885-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR