4-Bromo-2-ethylpyridine 98%

Supplier: Ambeed
Warning

A164863-250MG A164863-100MG A164863-1G
77028-778EA 141.1 USD
77028-778 77667-304 77667-306
4-Bromo-2-ethylpyridine 98%
4-Bromo-2-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD11223216
CAS Number: 156761-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR