1-Octadecanol 98%

Supplier: Ambeed
Warning

Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol

A164798-1000G A164798-100G A164798-10G A164798-25G A164798-500G
76997-974EA 33 USD
76997-974 76997-976 76997-978 76997-980 76997-982
1-Octadecanol 98%
1-Octadecanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
Boiling Pt: 336
MDL Number: MFCD00002823
CAS Number: 112-92-5
Merck Index: 12,08960

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR