5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine dihydrochloride 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A164701-1G A164701-100MG
76985-198EA 119.89 USD
76985-198 77683-356
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine dihydrochloride 97%, Ambeed.Inc
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine dihydrochloride

Precaución: For research use only.

Formula: C₅H₁₀Cl₂N₄
MW: 197.06 g/mol
MDL Number: MFCD23701368
CAS Number: 1429056-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR