2-Amino-5-bromo-N,N-dimethylbenzamide 98%

Supplier: Ambeed
Warning

A164568-1G A164568-250MG A164568-5G
77028-766EA 63.66 USD
77028-766 77028-768 77028-770
2-Amino-5-bromo-N,N-dimethylbenzamide 98%
2-Amino-5-bromo-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O
MW: 243.10 g/mol
MDL Number: MFCD09037777
CAS Number: 139253-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR