Methyl-6-bromo-1H-indazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-bromo-1H-indazole-4-carboxylate, 6-Bromo-4-indazolecarboxylic acid methyl ester

A164565-100G A164565-1G A164565-250MG A164565-25G A164565-5G
76997-958EA 600 USD
76997-958 76997-960 76997-962 76997-964 76997-966
Methyl-6-bromo-1H-indazole-4-carboxylate 97%
Methyl 6-bromo-1H-indazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MDL Number: MFCD07781319
CAS Number: 885518-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR