2-(3-(Trifluoromethyl)phenyl)morpholine hydrochloride 97%

Supplier: Ambeed
Warning

A164561-100MG A164561-250MG
77028-762EA 50.93 USD
77028-762 77028-764
2-(3-(Trifluoromethyl)phenyl)morpholine hydrochloride 97%
2-(3-(Trifluoromethyl)phenyl)morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClF₃NO
MW: 267.68 g/mol
MDL Number: MFCD01713452
CAS Number: 31599-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR