5-Bromo-4-methylpyridine-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A164415-10G
A164415-1G
A164415-250MG
A164415-25G
A164415-5G
A164415-100G
77011-352EA
116
USD
77011-352
77011-354
77011-356
77011-358
77011-360
77890-075
5-Bromo-4-methylpyridine-2-carboxylic acid 98%
5-Bromo-4-methylpyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₆BrNO₂ |
MDL Number:
MFCD07375100 CAS Number: 886365-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...