tert-Butyl (1-(4-chlorophenyl)-2-hydroxyethyl)carbamate 97%

Supplier: Ambeed
Warning

A164407-100MG A164407-1G A164407-250MG
77011-346EA 76 USD
77011-346 77011-348 77011-350
tert-Butyl (1-(4-chlorophenyl)-2-hydroxyethyl)carbamate 97%
tert-Butyl (1-(4-chlorophenyl)-2-hydroxyethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD24387088
CAS Number: 147353-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR