(R)-tert-Butyl (1-aminopropan-2-yl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-N-Boc-propane-1,2-diamine, tert-Butyl (R)-1-aminopropan-2-ylcarbamate

A164387-10G A164387-1G A164387-250MG A164387-5G
76997-948EA 215 USD
76997-948 76997-950 76997-952 76997-954
(R)-tert-Butyl (1-aminopropan-2-yl)carbamate 98%
(R)-tert-Butyl (1-aminopropan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD11112229
CAS Number: 100927-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR