4-(4-(2-Hydroxyethyl)piperazin-1-yl)butane-1-sulfonic acid 97%

Supplier: Ambeed
Warning

Synonyms: HEPBS

A164372-100G A164372-25G
76997-946EA 75.33 USD
76997-946 77671-320
4-(4-(2-Hydroxyethyl)piperazin-1-yl)butane-1-sulfonic acid 97%
4-(4-(2-Hydroxyethyl)piperazin-1-yl)butane-1-sulfonic acid

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂O₄S
MW: 266.35 g/mol
MDL Number: MFCD00677124
CAS Number: 161308-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR