4-((4-Chlorophenyl)sulfonyl)aniline 97%

Supplier: Ambeed
Warning

A164340-100MG A164340-1G A164340-250MG
76985-188EA 47.75 USD
76985-188 76985-190 76985-192
4-((4-Chlorophenyl)sulfonyl)aniline 97%
4-((4-Chlorophenyl)sulfonyl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO₂S
MW: 267.73 g/mol
MDL Number: MFCD00025350
CAS Number: 7146-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR