2-Bromo-5-chloro-1,3-dimethylbenzene 98%

Supplier: Ambeed
Warning

A164312-10G A164312-1G A164312-25G A164312-5G
77028-744EA 37.14 USD
77028-744 77028-746 77028-748 77028-750
2-Bromo-5-chloro-1,3-dimethylbenzene 98%
2-Bromo-5-chloro-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MDL Number: MFCD07780649
CAS Number: 103724-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR