(R)-Methyl-2-amino-3-(benzylthio)propanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Cys(Bzl)-OMe·HCl, H-Cys(Bzl)-OMe.HCl

A164311-1G A164311-25G A164311-5G A164311-10G
76997-932EA 7 USD
76997-932 76997-934 77600-320 77671-316
(R)-Methyl-2-amino-3-(benzylthio)propanoate hydrochloride 97%
(R)-Methyl-2-amino-3-(benzylthio)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClNO₂S
MDL Number: MFCD00034845
CAS Number: 16741-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR