Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate 98%

Supplier: Ambeed
Warning

Synonyms: Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid Stearyl Ester

A164280-100G A164280-25G A164280-10G
76997-930EA 11 USD
76997-930 77600-318 77671-314
Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate 98%
Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate

Precaución: for Research Use Only

Formula: C₃₅H₆₂O₃
MW: 530.88 g/mol
MDL Number: MFCD00017498
CAS Number: 2082-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR