3,5-Dibromo-2-chloro-4-pyridinamine 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-2-chloropyridin-4-amine

A164246-100MG A164246-1G A164246-250MG
77028-724EA 14.86 USD
77028-724 77028-726 77028-728
3,5-Dibromo-2-chloro-4-pyridinamine 95%
3,5-Dibromo-2-chloro-4-pyridinamine

Precaución: for Research Use Only

Formula: C₅H₃Br₂ClN₂
MW: 286.36 g/mol
MDL Number: MFCD11110699
CAS Number: 1054484-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR