2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

A164021-1G A164021-250MG A164021-25G A164021-5G
77011-334EA 50 USD
77011-334 77011-336 77011-338 77011-340
2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester 98%
2-(N-Methylaminomethyl)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₂
MDL Number: MFCD11855967
CAS Number: 1150271-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR