4-Amino-3-(methoxycarbonyl)benzoic acid 97%

Supplier: Ambeed
Warning

A163907-100MG A163907-1G
77011-332EA 71 USD
77011-332 77550-040
4-Amino-3-(methoxycarbonyl)benzoic acid 97%
4-Amino-3-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD11845550
CAS Number: 41684-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR