2-Propoxy-5-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (6-Propoxypyridin-3-yl)boronic acid

A163758-1G A163758-250MG A163758-5G
77028-712EA 83.82 USD
77028-712 77028-714 77028-716
2-Propoxy-5-pyridineboronic acid 98%
2-Propoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MW: 181.0000 g/mol
MDL Number: MFCD11855844
CAS Number: 1150114-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR