Methyl 2,4-dichloro-6-methylpyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A163747-10G A163747-250MG A163747-25G A163747-5G A163747-1G
76985-170EA 982.4 USD
76985-170 76985-172 76985-174 76985-176 77592-990
Methyl 2,4-dichloro-6-methylpyrimidine-5-carboxylate 95%
Methyl 2,4-dichloro-6-methylpyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MW: 221.04 g/mol
MDL Number: MFCD22572319
CAS Number: 36745-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR