1,4-Dibenzyl-L-aspartate-p-toluenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: H-Asp(OBzl)-Obzl.TosOH, L-Aspartic Acid Dibenzyl Ester p-Toluenesulphonate Salt, L-Aspartic acid dibenzyl ester p-toluenesulphonate salt Dibenzyl (2S)-2-aminobutanedioate 4-toluenesulphonate, H-Asp(OBzl)-OBzl.Tos-OH, L-Aspartic Acid 1,4-Dibenzyl Ester p-Toluenesulphonate, H-Asp(OBzl)-OBzl·TosOH, L-Aspartic acid dibenzyl ester p-toluenesulphonate

A163651-100G A163651-10G A163651-25G A163651-500G A163651-5G
76997-904EA 88 USD
76997-904 76997-906 76997-908 76997-910 76997-912
1,4-Dibenzyl-L-aspartate-p-toluenesulfonate 97%
1,4-Dibenzyl-L-aspartate-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₄
MDL Number: MFCD00065188
CAS Number: 2886-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR