1H-Pyrazolo[4,3-c]pyridine 98%
Supplier: Ambeed
|
Warning
|
A163636-100MG
A163636-250MG
A163636-5G
77028-706EA
18.04
USD
77028-706
77028-708
77028-710
1H-Pyrazolo[4,3-c]pyridine 98%
1H-Pyrazolo[4,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₆H₅N₃ |
MDL Number:
MFCD11846327 CAS Number: 271-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...