(3-Methyl-4-nitrophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Methyl-4-nitrophenyl)acetic acid

A163385-1G A163385-250MG
77011-318EA 60.48 USD
77011-318 77011-320
(3-Methyl-4-nitrophenyl)acetic acid 97%
(3-Methyl-4-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.18 g/mol
MDL Number: MFCD11036394
CAS Number: 143665-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR