3-(Hydroxymethyl)-5-nitrophenol 95%

Supplier: Ambeed
Warning

A163381-1G A163381-5G
76985-154EA 125 USD
76985-154 76985-156
3-(Hydroxymethyl)-5-nitrophenol 95%
3-(Hydroxymethyl)-5-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₇NO₄
MDL Number: MFCD11870181
CAS Number: 180628-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR