3,6-Bis(5-bromothiophen-2-yl)-2,5-bis(2-ethylhexyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%

Supplier: Ambeed
Warning

A163302-100MG A163302-1G A163302-250MG
76985-148EA 18 USD
76985-148 76985-150 76985-152
3,6-Bis(5-bromothiophen-2-yl)-2,5-bis(2-ethylhexyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%
3,6-Bis(5-bromothiophen-2-yl)-2,5-bis(2-ethylhexyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Precaución: for Research Use Only

Formula: C₃₀H₃₈Br₂N₂O₂S₂
MDL Number: MFCD16619382
CAS Number: 1000623-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR