3-Chloro-5-iodopyridin-4-amine 95%

Supplier: Ambeed
Warning

A163273-1G A163273-25G A163273-5G
77011-314EA 15 USD
77011-314 77011-316 77890-073
3-Chloro-5-iodopyridin-4-amine 95%
3-Chloro-5-iodopyridin-4-amine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
MDL Number: MFCD24450270
CAS Number: 1300750-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR