N-(2-Amino-4-fluorophenyl)methanesulfonamide 98%

Supplier: Ambeed
Warning

A163265-1G A163265-5G
77011-310EA 61.54 USD
77011-310 77011-312
N-(2-Amino-4-fluorophenyl)methanesulfonamide 98%
N-(2-Amino-4-fluorophenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₇H₉FN₂O₂S
MW: 204.22 g/mol
MDL Number: MFCD09047153
CAS Number: 156522-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR