Furo[3,2-b]pyridine 97%
Supplier: Ambeed
|
Warning
|
A163239-100MG
A163239-1G
A163239-250MG
A163239-5G
76985-140EA
35.01
USD
76985-140
76985-142
76985-144
76985-146
Furo[3,2-b]pyridine 97%
Furo[3,2-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅NO MW: 119.12 g/mol |
MDL Number:
MFCD11870744 CAS Number: 272-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...