5-Acetamido-2-chlorobenzoic acid 95%

Supplier: Ambeed
Warning

A163208-1G A163208-250MG A163208-5G
77011-308EA 33 USD
77011-308 77890-071 77890-072
5-Acetamido-2-chlorobenzoic acid 95%
5-Acetamido-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClNO₃
MDL Number: MFCD00466845
CAS Number: 719282-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR