2-Bromo-5-iodophenol 97%

Supplier: Ambeed
Warning

A163026-100MG A163026-1G A163026-250MG A163026-25G A163026-5G A163026-10G
77028-696EA 21.22 USD
77028-696 77028-698 77028-700 77592-982 77592-984 77600-308
2-Bromo-5-iodophenol 97%
2-Bromo-5-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
MW: 298.9000 g/mol
MDL Number: MFCD09907878
CAS Number: 932372-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR