2-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-6-trifluoromethylpyridine-4-boronic acid pinacol ester
A163005-100MG
A163005-1G
A163005-250MG
77011-290EA
39
USD
77011-290
77011-292
77011-294
2-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine 98%
2-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₂H₁₄BBrF₃NO₂ |
MDL Number:
MFCD17926520 CAS Number: 1256360-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...